missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA544184
Additional Details : Gewicht : 0.00010kg
Beschreibung
Peptide sequence: APRLLDFQKT KYARFMNHRV PAHKRYQPTE YEHAANCATH AFWIIPSILG Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%
MMD2 Polyclonal antibody specifically detects MMD2 in Human samples. It is validated for Western Blot.Spezifikation
MMD2 | |
Polyclonal | |
Unconjugated | |
MMD2 | |
4930518M15Rik; C88001; Mmd2; monocyte to macrophage differentiation associated 2; monocyte to macrophage differentiation factor 2; monocyte to macrophage differentiation-associated 2; monocyte-to-macrophage differentiation factor 2; monocyte-to-macrophage differentiation-associated protein 2; PAQR10; Progestin and adipoQ receptor family member 10; progestin and adipoQ receptor family member X | |
Rabbit | |
Affinity Chromatography | |
RUO | |
100505381, 221938 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8IY49 | |
MMD2 | |
Synthetic peptide directed towards the N-terminal of human MMD2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |