missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLX Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12042-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The product of this gene belongs to the family of basic helix-loop-helix leucine zipper (bHLH-Zip) transcription factors. These factors form heterodimers with Mad proteins and play a role in proliferation, determination and differentiation. This gene product may act to diversify Mad family function by its restricted association with a subset of the Mad family of transcriptional repressors, namely, Mad1 and Mad4. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.Spezifikation
MLX | |
Polyclonal | |
Unconjugated | |
MLX | |
bHLHd13, MAD7, Max like bHLHZip protein, MAX like protein X, MLX, MXD7, Protein BigMax, TCFL4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21428, 360631, 6945 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O08609, Q9UH92 | |
Mlx | |
MLX Fusion Protein Ag2671 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |