Learn More
Tocris Bioscience™ ML 240
GREENER_CHOICE
Artikelnummer. 18790789
Click to view available options
Menge:
10 mg
50 mg
Packungsgröße:
10 Milligramm
50 Milligramm
Artikelnummer. 18790789
Marke: Tocris Bioscience™ 5153/10
om dit product te kopen Registreer vandaag om een webaccount aan te maken
ATP-competitive inhibitor of p97 ATPase
ML 240 is an ATP-competitive inhibitor of p97 ATPase (VCP, IC50 = 110 nM). Exhibits antiproliferative activity in NCI-60 cancer cell lines and rapidly induces executioner caspases 3 and 7 in multiple colon cancer cells. Promotes accumulation of LC3-II and impairs autophagosome maturation. Also impairs the endoplasmic-reticulum-associated degradation (ERAD) pathway.
TRUSTED_SUSTAINABILITY
Spezifikation
Menge 10 mg
Produkttyp ML 240
Reinheit >99%
Molekulargewicht 396.44
Hemmstoffe ML 240
Nur für Forschungszwecke