Learn More
MKRN2OS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA564247
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: YDGRSDLHVG ITNTNGVVYN YSAHGVQRDG EGWEESISIP LLQPNMYGMM EQWDKYLEDF STSGAWLPHR YEDNH Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 73%.
MKRN2OS Polyclonal antibody specifically detects MKRN2OS in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
MKRN2OS | |
Polyclonal | |
Unconjugated | |
MKRN2OS | |
C3orf83; MKRN2 antisense gene protein 1; MKRN2 antisense RNA 1; MKRN2 antisense RNA 1 (non-protein coding); MKRN2 opposite strand; MKRN2 opposite strand protein; MKRN2-AS1; MKRN2OS | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100129480 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
H3BPM6 | |
MKRN2OS | |
Recombinant protein corresponding to Human MKRN2OS | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only