missing translation for 'onlineSavingsMsg'
Learn More

Mitomycin C

Artikelnummer. 15425459
Ansicht ändern
Click to view available options
Menge:
5 mg
10 mg
Packungsgröße:
10 Milligramm
5 Milligramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
15425459 10 mg 10 Milligramm
15415459 5 mg 5 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15425459 Lieferant Thermo Scientific Alfa Aesar Lieferanten-Nr. J63193.MA
 Eingeschränkter Artikel
Lager vorrätig
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Mitomycin C, CAS # 50-07-7, is a methylazirinopyrroloindoledione compound showing antineoplastic and antibiotic activities. It is active against a broad-spectrum of gram-positive, gram-negative, and acid-fast bacilli bacteria.

Mitomycin C is used as a chemotherapeutic agent due to its antitumor activity. It is used to treat upper gastro-intestinal cancers. It serves as an antineoplastic agent which inhibits DNA synthesis and induces apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Mitomycin C is a methylazirinopyrroloindoledione compound isolated from the bacterium Streptomyces caespitosus, and other Streptomyces bacterial species that has antineoplastic and antibiotic activities
• This compound acts as an inhibitor of DNA synthesis, nuclear division, as well as an apoptosis inducer. It can create oxygen radicals, and produce inter-strand DNA crosslinks, inhibiting DNA synthesis
• Mitomycin C shows alkylating activity and mainly targets the guanine nucleoside sequence 5′-CpG-3′. It inhibits DNA synthesis by covalently reacting with DNA, forming crosslinks between complementary strands of DNA. Consequently, this interaction prevents the separation of complementary DNA strands, inhibiting DNA replication

Application

• Mitomycin C is active against a broad-spectrum of gram-positive, gram-negative, and acid-fast bacilli bacteria
• Mitomycin C has strong antineoplastic activity, mainly against Ehrlich ascites tumor cell lines
• This compound can inhibit B and T cells and macrophage proliferation. It can also impair antigen presentation, as well as the secretion of interferon-γ, TNF- α, and IL-2
• This compound is used to generate mitotically inactive feeder cells in cell culture systems, such as the mitotically inactive fibroblasts used in embryonic stem cell systems
• It has been used to treat basal-like cancer cell line in vitro experiments
• It is particularly toxic to hypoxic cells inhibiting RNA and protein synthesis at high concentrations

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 50-07-7
Summenformel C15H18N4O5
Molekulargewicht (g/mol) 334.332
MDL-Nummer MFCD00078109
InChI-Schlüssel NWIBSHFKIJFRCO-WUDYKRTCSA-N
PubChem CID 5746
ChEBI CHEBI:27504
SMILES CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N

Spezifikation

Chemischer Name oder Material Mitomycin C
Typ Mitomycin C
Physikalische Form Pulver
Schmelzpunkt 360°C
Farbe Blau bis Violett
Menge 10 mg
UN-Nummer UN2811
Beilstein 7231816
Merck Index 14,6215
Löslichkeitsinformationen Soluble in methanol,water and dimethyl sulfoxide.
Formelmasse 334.33
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • Mitomycin C
Signalwort
  • Gefahr
Gefahrenkategorie
  • Akute Toxizität Kategorie 3
  • Karzinogenität Kategorie 2
Gefahrenhinweis
  • H301-Giftig bei Verschlucken.
  • H351-Kann vermutlich Krebs erzeugen.
Sicherheitshinweis
  • P201-Vor Gebrauch besondere Anweisungen einholen.
  • P264-Nach Gebrauch gründlich waschen.
  • P280-Schutzhandschuhe/Schutzkleidung/Augenschutz/Gesichtsschutz tragen.
  • P301+P330+P331-BEI VERSCHLUCKEN: Mund ausspülen. KEIN Erbrechen herbeiführen.
  • P308+P313-BEI Exposition oder falls betroffen: Ärztlichen Rat einholen/ärztliche Hilfe hinzuziehen.
  • P405-Unter Verschluss aufbewahren.
Ergänzende Informationen
  • MIXTURE LIST-Contain: Mitomycin-C

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.