missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Mirabegron

Click to view available options
Menge:
5 mg
25 mg
Packungsgröße:
25 Milligramm
5 Milligramm
Description
Mirabegron is a potent and selective β3-adrenoceptor agonist (EC50 = 22.4 nM). Displays >440-fold selectivity for β3 over β1 and β2 adrenoceptors. Increases cAMP concentration in CHO cells expressing human β3-adrenoceptor. Displays relaxant effect in ex vivo rat bladder muscle preparation precontracted with carbachol (Cat. No. 2810). Decreases frequency of bladder contraction in rats. Also promotes browning of white adipose tissue, reduces body weight, and improves glucose tolerance and insulin sensitivity in mice fed a high fat diet.
Specifications
Specifications
| Chemischer Name oder Material | 2-Amino-N-[4-[2-[[(2R)-2-hydroxy-2-phenylethyl]amino]ethyl]phenyl]-4-thiazoleacetamide |
| CAS | 223673-61-8 |
| Menge | 25 mg |
| Summenformel | C21H24N4O2S |
| Reinheit | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction