Beschreibung
- Extremely compact footprint
- Acceleration and deceleration time <13 seconds
- User-friendly digital display for time and speed
- Metal rotor bowl
- Separate short-spin key for fast and comfortable centrifugation
- Optimal ventilation flow reduces heating and protects temperature-sensitive samples
- Soft-touch lid closure for ergonomic lid locking
- Automatic lid opening at the end of the run to prevent sample warming and to allow easy access to samples
Spezifikation
Spezifikation
| Fassungsvermögen | 12 x 1.5/2 mL Tubes |
| Max. Geschwindigkeit | 14,500 rpm |
| Max. RZB | 14,100 x g |
| Gekühlt | No |
| Produkttyp | Mini centrifuge, IVD Switzerland Version |