Learn More
Millipore™ Milliflex-Inkubationskassette
Used for the recovery of a broad range of heterotrophic microorganisms such as common aerobic and facultative anaerobic bacteria found in water. Millipore™ Milliflex Incubation Cassette is prefilled with tryptic soy agar.
Marke:  Millipore™ MXSMCTS48
Additional Details : Gewicht : 0.10000kg
Artikelnummer. 10793605
  • 325.00 CHF / 48 Stück
Verfügbar ab
12-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • Combines products, accessories and services to immediately streamline your microbial contamination testing workflow.
  • Leverages your quality assurance level by avoiding risks of exogenous contamination and ensure compliance to all GMP/GLP requirements.
Order Info
ShelfLife
Spezifikation
Spezifikation
120 h
10 mL
Milliflex Cassette
Trypton-Soja-Agar
E. coli , A. niger, C. albicans, B. subtilis, S. aureus, P. aeruginosa
Zur Behebung von Belastungen mit Keimzahlen heterotropher Bakterien, die in verschiedenen Gewässerarten vorkommen, insbesondere in hochreinem Wasser
Videos