missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MICA Mouse anti-Human, Clone: 1E2C8, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66384-1-IG-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
MICA is the higly polymorphic MHC (HLA) class I chain-related gene A. The protein product is expressed on the cell surface, although unlike canonical class I molecules does not seem to associate with beta-2-microglobulin. It is thought that MICA functions as a stress-induced antigen that is broadly recognized by intestinal epithelial gamma delta T cells.Spezifikation
MICA | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q29983 | |
MICA | |
MICA Fusion Protein Ag3298 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1E2C8 | |
Unconjugated | |
MICA | |
MIC A, MICA, PERB11.1 | |
Mouse | |
Protein G | |
RUO | |
100507436 | |
-20°C | |
Liquid |