Learn More
Invitrogen™ MIA-2 Human ELISA Development Kit (ABTS), PeproTech®, PeproTech®
ELISA
Marke: Invitrogen™ 900-K357
Additional Details : Gewicht : 0.03500kg
Beschreibung
CTAGE5/MIA-2 is a secreted cytokine, and a member of the MIA/OTOR family. Members of this family, which also includes MIA, OTOR, and TANGO, share a Src homology-3 (SH3)-like domain. MIA-2 is predominantly expressed in hepatocytes. Elevated levels of MIA-2 may represent a clinically useful marker for diagnosis of hepatic disease activity and severity.Spezifikation
Q96PC5 | |
4253 | |
10 x 96 Tests | |
Refer to product documentation for component specific storage temperature. |
ELISA | |
Mia2 | |
RUO | |
Human |