Learn More
Motic™ MHV-2-RS Dual-Head Observation Optical Bridge, BA210/BA310
Multi-head viewing system suitable for BA210 and BA310 Microscopes. Motic™ MHV-2-RS Dual-Head Observation Optical Bridge, BA210/BA310 facilitates viewing by up to 2 individuals.
Marke: Motic™ 1101000301471
Additional Details : Gewicht : 1.00000kg
Beschreibung
- Right-side unit.
- Side-by-side orientation.
- LED pointer on primary unit.
- Adjustable height support apparatus for secondary unit (1 piece).
- Additional head and eyepieces required.