Learn More
Motic™ MHV-2-RS Dual-Head Observation Optical Bridge, BA210/BA310
Multi-head viewing system suitable for BA210 and BA310 Microscopes. Motic™ MHV-2-RS Dual-Head Observation Optical Bridge, BA210/BA310 facilitates viewing by up to 2 individuals.
Marke:  Motic™ 1101000301471
Additional Details : Gewicht : 1.00000kg
Artikelnummer. 15753568
  • 22.95 CHF / 1 Stück
Verfügbar ab: 12-07-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • Right-side unit.
  • Side-by-side orientation.
  • LED pointer on primary unit.
  • Adjustable height support apparatus for secondary unit (1 piece).
  • Additional head and eyepieces required.