missing translation for 'onlineSavingsMsg'
Learn More

MHC Class II (I-A) Monoclonal Antibody (NIMR-4), FITC, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Marke:  Affymetrix eBioscience 11-5322-82

152.00 CHF gültig bis 2024-03-29
Benutzen Sie den Promotions-Code "21615" um Ihren Promotions-Preis zu erhalten.



Additional Details : Gewicht : 0.25000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15228239

  • 228.00 CHF / 100 Mikrogramm
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Description: The monoclonal antibody NIMR-4 reacts specifically with a non-polymorphic I-A-encoded epitope on MHC Class II antigens. Class II antigens are predominantly expressed on antigen-presenting cells including B lymphocytes, macrophages, dendritic cells and certain epithelial cells. MHC Class II molecules are important for the selection of the TCR repertoire during the processes of positive and negative selection. MHC Class II expression is required for the positive selection of CD4+ T cells. Applications Reported: This NIMR-4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This NIMR-4 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

MHC class II molecules are heterodimers of non-covalently associated alpha (31-34 kDa) and beta (26-29 kDa) chains. Major histocompatibility complex class II antigen presentation requires the participation of lysosomal proteases in two convergent processes. First, the antigens endocytosed by the antigen presenting cells must be broken down into antigenic peptides. Second, class II molecules are synthesized with their peptide-binding site blocked by invariant chain (Ii), and they aquire the capacity to bind antigens only after Ii has been degraded in the compartments where peptides reside. MHC class II molecules present exogenously derived antigen to CD4+ T lymphocytes, which are usually T helper cells. CD4 interacts with non-polymorphic residues of MHC class II.
Spezifikation

Spezifikation

MHC Class II (I-A)
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P01921, P06342, P06343, P06344, P06345, P06346, P14483
H2-Ab1
Affinity chromatography
RUO
14961
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
NIMR-4
FITC
H2-Ab1
Abeta; AI845868; H-2 class II histocompatibility antigen, A beta chain; H-2Ab; H2-Ab; H2-Ab1; H2-iabeta; histocompatibility 2, class II antigen A, beta 1; Ia2; Ia-2; IAb; I-Abeta; major histocompatibility complex class II beta chain; MHC class II antigen A beta; MHC class II H2-IA-beta-psi; response to metastatic cancers 1; Rmcs1
Rat
100 μg
Primary
Mouse
Antibody
IgG2b
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.