missing translation for 'onlineSavingsMsg'
Learn More

MHC Class II (I-A/I-E) Rat anti-Mouse, Super Bright 436, Clone: M5/114.15.2, eBioscience™

Rat Monoclonal Antibody

Marke:  Invitrogen 62-5321-80

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983424

  • 141.00 CHF / 25 Mikrogramm


Verfügbar ab
08-10-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

When using two or more Super Bright dye- conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product No. SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. MHC (major histocompatibility complex) class II molecules are transmembrane glycoproteins expressed on the surface of professional antigen-presenting cells, such as macrophages, dendritic cells and B cells. Before their exposition on the cell surface, the MHC class II molecules react with endocytosed exogenous antigens, which are then presented to the T cells. The antigen-binding grove between MHC class II alpha and beta chain is open at both ends and is 15-24 amino acid residues long.
Spezifikation

Spezifikation

MHC Class II (I-A/I-E)
Monoclonal
0.2 mg/mL
PBS with BSA and 0.09% sodium azide; pH 7.2
P06342
H2-Ab1
IgG2b, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
14961
Flow Cytometry
M5/114.15.2
Super Bright 436
H2-Ab1
Liquid
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Videos
Dokumentation