missing translation for 'onlineSavingsMsg'
Learn More

MGC50811 Recombinant Protein, Abnova

Marke:  Abnova H00375307-P01.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16150394

  • 458.00 CHF / 25 Mikrogramm


Verfügbar ab
01-07-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Spezifikation

Spezifikation

AAH52750
Solution
375307
MGC50811 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C2orf62
Human
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
68.31
8
Glutathione Sepharose 4 Fast Flow
25 ug
MSSKVYSTGSRAKDHQPSGPECLPLPEANAEAIDFLSSLHKEELQMLFFSETLAMVSDTGEPQGELTIEVQRGKYQEKLGMLTYCLFVHASSRGFLDKMLCGNSLLGYLSEKLELMEQHSQDFIKFLILPMERKMSLLKQDDQLAVTRSIKEGEEVKTGVTSFPWSSIKGFISEAANLVLLRVMAWRRMVPSNARFLTLDTEGKLCYLTYQNLGFQTIQVDHQQAEVFIVEQTVHAEEGIPMSCQYYLLSDGHLAKRIQVGSPGCCIITKMPILREEDEIEPRPVFEKKPLVWEEDMELYSKFLDRKEELRLGHASYLRQHPEAHALISDFLLFLLLRQPEDVVTFAAEFFGPFDPWRPSSPALGSSHRPNPFRSLEPEGDARSGAA
MGC50811
C2orf62
Wheat Germ (in vitro)
GST
Videos
Dokumentation