Learn More
LV-11 Automatic Feeder for AG, PG-S and PR Series Balances
Alle ansehen Mettler Toledo ProdukteDescription
Includes
Comes with feed chute, LocalCAN interface cable, power cable and operating instructions.
The LV-11 Feeder delivers small samples with diameters ranging from 1 to 33mm—ideal for pharmaceuticals, as well as tiny electronic or mechanical parts. A photocell automatically monitors each item as it passes by and interrupts the feed after each one—restarting after the balance has validated the weight. And, if needed, the feeder will empty itself when a sample of a desired size has been reached.Selectable four-step vibration intensity (feed rate) can be set to match the size, surface texture or shape of items. LED on front of feeder clearly indicates the intensity level.To create a fully automatic system that provides complete verification of your process, simply team up the LV-11 Feeder with one of the following: an AG or PG-S Series balance (outfitted with LC-G LocalCAN interface, 01-918-91) and the SQC14 Quality Control System1 (09-316-33); a PR Series balance and the SQC15 Quality Control Systems (01-913-77N, 01-913-77P); or a computer1. The built-in bidirectional RS-232-C and LocalCAN interfaces ensure error-free data transmission between your computer or weighing system. For 115/220V 50/60Hz, 15VA, switch-selectable.
Informations sur la commande
Overall dimensions: 8 7/8L x 8 7/8W x 10 1/4 in.H (22.5 x 22.5 x 26cm). Net weight 25 3/8 lb. (11.5kg).
Spécification
Spécification
| Zur Verwendung mit (Geräte) | XP Precision Balance |
| Zur Verwendung mit (Anwendung) | Small items |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.