missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mono-Methyl-Histone H1.2 (Lys186) Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA5112456
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Mono-Methyl-Histone H1.2 (Lys186) Polyclonal Antibody for Western Blot, ICC/IF, ELISA, ChIP
Histones are basic nuclear proteins responsible for nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H1 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
Spezifikation
| Mono-Methyl-Histone H1.2 (Lys186) | |
| Polyclonal | |
| Unconjugated | |
| H1-2 | |
| 0610008C09Rik; CTL-1; H1 histone family, member 2; H1 VAR.1; H1.2; H1.2 linker histone, cluster member; H1-2; H1C; H1f2; H1s-1; H1var1; His1a; Hist1h1c; histone 1, H1c; histone cluster 1, H1c; histone H1; histone H1.1; histone H1.2; Histone H1c; histone H1d; Histone H1s-1; LOC684681; MGC3992; similar to Histone H1.2 (H1 VAR.1) (H1c) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3006 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ChIP Assay, ELISA, Western Blot, Immunocytochemistry | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P16403 | |
| H1-2, HIST1H1C | |
| Peptide sequence around site of Mono-methyl-Lys (186) derived from Human Histone H1.2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur