missing translation for 'onlineSavingsMsg'
Learn More

Methyl-4-Brompyrrol-2-Carboxylat, 97 %, Thermo Scientific Chemicals

Catalog Number 15426807
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
934-05-4
C6H6BrNO2
204.023
MFCD00832855
ZXFCRVGOHJHZNF-UHFFFAOYSA-N
methyl 4-bromopyrrole-2-carboxylate, 4-bromo-2-methoxycarbonyl-1h-pyrrole, 4-bromo-1h-pyrrole-2-carboxylic acid methyl ester, methyl4-bromo-1h-pyrrole-2-carboxylate, 4-bromopyrrole-2-carboxylic acid methyl ester, 1h-pyrrole-2-carboxylic acid, 4-bromo-, methyl ester, pubchem13045, acmc-209rlm, ksc497g1p, 4-bromo-2-methoxycarbonylpyrrole
2763594
Methyl-4-brom-1H-pyrrol-2-carboxylat
COC(=O)C1=CC(=CN1)Br
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
934-05-4
C6H6BrNO2
204.023
MFCD00832855
ZXFCRVGOHJHZNF-UHFFFAOYSA-N
methyl 4-bromopyrrole-2-carboxylate, 4-bromo-2-methoxycarbonyl-1h-pyrrole, 4-bromo-1h-pyrrole-2-carboxylic acid methyl ester, methyl4-bromo-1h-pyrrole-2-carboxylate, 4-bromopyrrole-2-carboxylic acid methyl ester, 1h-pyrrole-2-carboxylic acid, 4-bromo-, methyl ester, pubchem13045, acmc-209rlm, ksc497g1p, 4-bromo-2-methoxycarbonylpyrrole
2763594
Methyl-4-brom-1H-pyrrol-2-carboxylat
COC(=O)C1=CC(=CN1)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Methyl 4-bromopyrrole-2-carboxylate
Schmelzpunkt 100°C to 102°C
Menge 1 g
Formelmasse 204
Reinheit (%) 97%

RUO – Nur für Forschungszwecke

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'