missing translation for 'onlineSavingsMsg'
Learn More

Methyl2-Amino-5-Methyl-4-Phenylthiophen-3-Carboxylat, 97%, Thermo Scientific™

Catalog Number p-7032106
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
350988-88-4
C13H13NO2S
247.31
MFCD01922145
AIHYNLWSPTZXGT-UHFFFAOYSA-N
2-amino-5-methyl-4-phenyl-thiophene-3-carboxylic acid methyl ester, methyl 2-amino-5-methyl-4-phenyl-3-thiophenecarboxylate, 2-amino-5-methyl-4-phenylthiophene-3-carboxylic acid methyl ester, cbmicro_030702, 3-thiophenecarboxylicacid, 2-amino-5-methyl-4-phenyl-, methyl ester
734155
methyl 2-amino-5-methyl-4-phenylthiophene-3-carboxylate
COC(=O)C1=C(N)SC(C)=C1C1=CC=CC=C1
This item is not returnable. View return policy
350988-88-4
C13H13NO2S
247.31
MFCD01922145
AIHYNLWSPTZXGT-UHFFFAOYSA-N
2-amino-5-methyl-4-phenyl-thiophene-3-carboxylic acid methyl ester, methyl 2-amino-5-methyl-4-phenyl-3-thiophenecarboxylate, 2-amino-5-methyl-4-phenylthiophene-3-carboxylic acid methyl ester, cbmicro_030702, 3-thiophenecarboxylicacid, 2-amino-5-methyl-4-phenyl-, methyl ester
734155
methyl 2-amino-5-methyl-4-phenylthiophene-3-carboxylate
COC(=O)C1=C(N)SC(C)=C1C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Methyl 2-amino-5-methyl-4-phenylthiophene-3-carboxylate
Schmelzpunkt 113°C to 114°C
Menge 1 g
Formelmasse 247.32
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'