missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,4-difluorbenzoat, 98 %, Thermo Scientific™

Catalog Number p-7031182
Click to view available options
Menge:
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
106614-28-2
C8H6F2O2
172.131
MFCD03093799
YQUHULOBTDYMAG-UHFFFAOYSA-N
2,4-difluoro-benzoic acid methyl ester, 2,4-difluorobenzoic acid methyl ester, methyl-2,4-difluorobenzoate, methyl 2,4-difluoro benzoate, benzoic acid, 2,4-difluoro-, methyl ester, pubchem15678, acmc-1c4sx, methyl 2,4-diflourobenzoate, ksc494g3p, rarechem al bf 0213
2782897
Methyl2,4-difluorbenzoat
COC(=O)C1=C(C=C(C=C1)F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
106614-28-2
C8H6F2O2
172.131
MFCD03093799
YQUHULOBTDYMAG-UHFFFAOYSA-N
2,4-difluoro-benzoic acid methyl ester, 2,4-difluorobenzoic acid methyl ester, methyl-2,4-difluorobenzoate, methyl 2,4-difluoro benzoate, benzoic acid, 2,4-difluoro-, methyl ester, pubchem15678, acmc-1c4sx, methyl 2,4-diflourobenzoate, ksc494g3p, rarechem al bf 0213
2782897
Methyl2,4-difluorbenzoat
COC(=O)C1=C(C=C(C=C1)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Methyl 2,4-difluorobenzoate
Menge 5 g
Formelmasse 172.13
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'