missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perthan in Methanol 100μ g/ml, Fisher Chemical™
Click to view available options
Menge:
1 ml
Verpackung:
Ampulle
Spezifikation
Konzentration | 100 μg/ml |
CAS | 72-56-0 |
Verpackung | Ampulle |
Menge | 1 ml |
Summenformel | C18H20Cl2 |
InChI-Schlüssel | QFMDFTQOJHFVNR-UHFFFAOYSA-N |
SMILES | CCC1 = CC = C (C = C1) C (C2 = CC = C (C = C2) CC) C (Cl) Cl |
IUPAC-Name | 1-[2,2-Dichloro-1-(4-ethylphenyl)ethyl]-4-ethylbenzol |
Molekulargewicht (g/mol) | 307.258 |
PubChem CID | 6295 |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'