missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MERS Coronavirus ORF4a Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA5112044
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
MERS Coronavirus ORF4a Polyclonal Antibody for Western Blot
Middle East respiratory syndrome-related coronavirus (MERS), or EMC/2012, is a species of coronavirus which infects humans, bats, and camels. The infecting virus is an enveloped, positive-sense, single-stranded RNA virus which enters its host cell by binding to the DPP4 receptor. The absence of all four Middle East respiratory syndrome CoV (MERS-CoV) accessory ORFs (deletion of ORF3, -4a, -4b, and -5 [dORF3-5]) has major implications for viral replication and pathogenesis.
Spezifikation
| MERS Coronavirus ORF4a | |
| Polyclonal | |
| Unconjugated | |
| MERS; Middle East Respiratory Syndrome | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of MERS-CoV ORF4a (MERS-CoV (strain United Kingdom/H123990006/2012)). The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.05 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur