Learn More
Trajan Scientific and Medical™ MEPS BINs for XCHANGE/eVol Syringes
Miniaturization of conventional sample preparation devices. Trajan Scientific and Medical MEPS BINs for XCHANGE/eVol Syringes are micro extraction by packed sorbent.
Marke:  Trajan Scientific and Medical™ 2900712
Additional Details : Gewicht : 0.01000kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 15229253
  • 605.00 CHF / 5 Stück
Verfügbar ab
14-08-2020
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • Save hours in sample preparation
  • Reduce solvent consumption and sample volume
  • Available in reversed phase, normal phase, mixed mode and ion exchange chemistries
Spezifikation
Spezifikation
C8
MEPS Barrel Insert and Needle (BIN)
Cone Tipped Needle
XCHANGE/eVol Syringes