missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEK6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | MEK6 |
---|---|
Konzentration | 0.21 mg/mL |
Anwendungen | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16850624
|
Proteintech
12745-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 13-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16840624
|
Proteintech
12745-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 13-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This gene encodes a member of the dual specificity protein kinase family, which functions as a mitogen-activated protein (MAP) kinase kinase. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals. This protein phosphorylates and activates p38 MAP kinase in response to inflammatory cytokines or environmental stress. As an essential component of p38 MAP kinase mediated signal transduction pathway, this gene is involved in many cellular processes such as stress induced cell cycle arrest, transcription activation and apoptosis.Spezifikation
MEK6 | |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
P52564, P70236 | |
114495, 26399, 5608 | |
MEK6 Fusion Protein Ag3706 | |
Primary | |
-20°C | |
MAP2K6 |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
MAP kinase kinase 6, MAP2K6, MAPK/ERK kinase 6, MAPKK 6, MAPKK6, MEK 6, MEK6, MKK6, PRKMK6, SAPKK3 | |
Map2k6 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |