Learn More
Abnova™ MED31 Recombinant Protein
Marke:  Abnova™ H00051003-P01.25ug
Additional Details : Gewicht : 0.00010kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 16129492
  • 657.00 CHF / 25 Mikrogramm
Verfügbar ab: 13-07-2022
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spezifikation
Spezifikation
AAH12539
Solution
51003
MED31 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MED31
Human
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
40.15
8
Glutathione Sepharose 4 Fast Flow
25 ug
MAAAVAMETDDAGNRLRFQLELEFVQCLANPNYLNFLAQRGYFKDKAFVNYLKYLLYWKDPEYAKYLKYPQCLHMLELLQYEHFRKELVNAQCAKFIDEQQILHWQHYSRKRMRLQQALAEQQQQNNTSGK
3110004H13Rik/CGI-125/FLJ27436/FLJ36714/Soh1
MED31