missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20028-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This antibody is specific to MED12.
The initiation of transcription is controlled in part by a large protein assembly known as the preinitiation complex. A component of this preinitiation complex is a 1.2 MDa protein aggregate called Mediator. This Mediator component binds with a CDK8 subcomplex which contains the protein encoded by this gene, mediator complex subunit 12 (MED12), along with MED13, CDK8 kinase, and cyclin C.Spezifikation
MED12 | |
Polyclonal | |
Unconjugated | |
MED12 | |
ARC240, CAG repeat protein 45, CAGH45, FGS1, HOPA, KIAA0192, MED12, mediator complex subunit 12, OKS, OPA containing protein, OPA1, TNRC11, TRAP230 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
59024, 679693, 9968 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A2AGH6, Q93074 | |
Med12 | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |