Learn More
MCT1 Rabbit anti-Human, Polyclonal, Proteintech
Artikelnummer. 16899634 Alle ansehen Proteintech Produkte
Change view
Click to view available options
Menge:
20 μL
150 μL
Packungsgröße:
150 Mikroliter
20 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16899634 20 μL 20 Mikroliter
16889634 150 μL 150 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16899634 Lieferant Proteintech Lieferanten-Nr. 201391AP20UL
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Rabbit Polyclonal Antibody
SLC16A1 is a monocarboxylate transporter (MCT1) that mediates the movement of lactate and pyruvate across cell membranes. Import and export of these substrates by tissues such as erythrocytes, muscle, intestine, and kidney are ascribed largely to the action of a proton-coupled MCT.
Spezifikation
Antigen MCT1
Anwendungen Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunoprecipitation
Klassifikation Polyclonal
Konzentration 0.29 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gen SLC16A1
Gen-Zugriffsnummer P53985
Gen-Alias MCT, MCT 1, MCT1, Monocarboxylate transporter 1, SLC16A1
Gensymbole SLC16A1
Wirtsspezies Rabbit
Immunogen MCT1 Fusion Protein Ag14098
Reinigungsverfahren Antigen Affinity Chromatography
Menge 20 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 6566
Zielspezies Human
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG