missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MATK (Hyl) Recombinant Human Protein
MATK (Hyl) Recombinant Human Protein
Marke: Invitrogen PV3370
Additional Details : Gewicht : 0.05250kg
Beschreibung
MATK (Hyl) is a non-receptor CSK-type protein tyrosine kinase that is thought to function in signal transduction pathways important in megakaryocyte growth and⁄or differentiation.Spezifikation
NP_647611 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
53kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
MATK (Hyl), CTK | |
MATK was pre-diluted in enzyme dilution buffer and assayed in 50mM Tris (pH 7.5), 10mM MgCl2, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C | |
MATK (Hyl) Recombinant Human Protein | |
Human | |
E. coli | |
Cytoplasmic tyrosine kinase |
0.4mg/mL | |
4145 | |
MATK (Hyl) Recombinant Human Protein | |
–80°C | |
1 Each | |
MATK (Hyl) Recombinant Human Protein | |
CSK | |
490nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 4μg/mL | |
His Tag | |
Kinase and inhibitors | |
90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.