missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MAT2B Recombinant Protein
Marke: Abnova™ H00027430-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spezifikation
AAH05218 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
61.27 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MPEMPEDMEQEEVNIPNRRVLVTGATGLLGRAVHKEFQQNNWHAVGCGFRRARPKFEQVNLLDSNAVHHIIHDFQPHVIVHCAAERRPDVVENQPDAASQLNVDASGNLAKEAAAVGAFLIYISSDYVFDGTNPPYREEDIPAPLNLYGKTKLDGEKAVLENNLGAAVLRIPILYGEVEKLEESAVTVMFDKVQFSNKSANMDHWQQRFPTHVKDVATVCRQLAEKRMLDPSIKGTFHWSGNEQMTKYEMACAIADAFNLPSSHLRPITDSPVLGAQRPRNTQLDCSKLETLGIGQRTPFRIGIKESLWPFLIDKRWRQTVFH | |
MAT-II/MATIIbeta/MGC12237/Nbla02999/SDR23E1/TGR | |
MAT2B | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
27430 | |
MAT2B (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MAT2B | |
Human | |
Recombinant | |
Solution |