Learn More
Aalborg™ Aluminum Digital Mass Flowmeter
Click to view available options
Typ:
Massendurchflussmesser
Packungsgröße:
1 Stück
Beschreibung
- Supports up to 23 Engineering Units (including User Defined).
- Stores calibration data for up to 10 gases.
- Programmable Totalizer indicates total gas quantity.
- High and low gas flow Alarm limits with preset delay interval.
- Two sets of user-programmable electromechanical SPDT relays with latch option.
- User-selectable analog 0-5 Vdc or 4-20mA outputs.
- Internal Conversion factors for up to 32 gases.
- Digital Interface (RS-232 / RS-485, Profibus DP available).
- Multi-Drop Capability of up to 256 units (RS-485 option).
- Optional Profibus DP interface with I&M functionality.
- Automatic sensor zero offset adjustment (via digital interface or local push button).
- Self-Diagnostic Tests.
- Local 2 x 16 characters LCD display with adjustable back light (optional).
Spezifikation
Spezifikation
| Typ | Massendurchflussmesser |
| Flussratenbereich | 2 LPM |