missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MAPK10 (JNK3), GST Tag Recombinant Protein
Human MAPK10 (JNK3), GST Tag Recombinant Protein
Marke: Invitrogen PV4563
Additional Details : Gewicht : 0.50000kg
Beschreibung
MAPK10 (JNK3) is a neuronal-specific form of c-Jun N-terminal kinases (JNKs). MAPK10 is involved in neuronal apoptosis and phosphorylates MAP2K4.Spezifikation
NP_002744 | |
76.6 kDa | |
MAPK10 (JNK3) | |
Human | |
GST Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic | |
Ser⁄Thr Kinases: CMGC Group | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
MAPK10 | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |
Kinase Assay | |
10 μg | |
MAP Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
10 μg | |
Dry Ice | |
N-Terminal | |
mitogen-activated protein kinase 10 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |