missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MAPK10 (JNK3), GST Tag Recombinant Protein
Human MAPK10 (JNK3), GST Tag Recombinant Protein
838.00 CHF
Spezifikation
Zugriffsnummer | NP_002744 |
---|---|
Zur Verwendung mit (Anwendung) | Kinase Assay |
Molekulargewicht | 76.6 kDa |
Gen-Alias | MAPK10 (JNK3) |
Kinase-Familie | MAP Kinase Family |
Beschreibung
MAPK10 (JNK3) is a neuronal-specific form of c-Jun N-terminal kinases (JNKs). MAPK10 is involved in neuronal apoptosis and phosphorylates MAP2K4.Spezifikation
NP_002744 | |
76.6 kDa | |
MAP Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors |
Kinase Assay | |
MAPK10 (JNK3) | |
Human | |
GST Tag | |
Ser⁄Thr Kinases: CMGC Group | |
Baculovirus |