Learn More
MAP9 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 26078-1-AP-150UL
 Lernen Sie weitere Varianten dieses
Artikelnummer. 16854605
  • 385.92 CHF / 150 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
ASAP is a microtubule-associated protein required for spindle function, mitotic progression, and cytokinesis (Saffin et al., 2005 ).
Spezifikation
Spezifikation
MAP9
Polyclonal
Unconjugated
MAP9
ASAP, Aster associated protein, MAP9
Rabbit
Antigen Affinity Chromatography
RUO
79884
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.47 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q49MG5
MAP9
MAP9 Fusion Protein Ag23446
150 μL