missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP3K3 (MEKK3) Recombinant Human Protein
MAP3K3 (MEKK3) Recombinant Human Protein
Marke: Invitrogen PV3876
Additional Details : Gewicht : 0.05250kg
Beschreibung
MAP3K3 (MEKK3) directly regulates MAPK8 and extracellular signal-regulated protein kinase (ERK) pathways by phosphorylating multiple MAP2Ks.Spezifikation
NP_002392 | |
Cell Viability, Pharma and Biopharma, Cell Proliferation and Function, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
99.8kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
MAP3K3 (MEKK3) | |
MAP3K3 (MEKK3) was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 100μM ATP, 500μg/mL myelin basic protein (MBP) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
MAP3K3 (MEKK3) Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.21mg/mL | |
114294, 4215 | |
MAP3K3 (MEKK3) Recombinant Human Protein | |
–80°C | |
1 Each | |
MAP3K3 (MEKK3) Recombinant Human Protein | |
STE11 | |
34nmoles of phosphate transferred to myelin basic protein (MBP) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 8.33μg/mL | |
GST Tag | |
Kinase and inhibitors | |
60% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.