missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP3K1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19970-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The antibody recognizes the N-term of MAP3K1.
Mitogen-activated protein kinase (MAPK) signaling cascades include MAPK or extracellular signal-regulated kinase (ERK), MAPK kinase (MKK or MEK), and MAPK kinase kinase (MAPKKK or MEKK). MAPKK kinase/MEKK phosphorylates and activates its downstream protein kinase, MAPK kinase/MEK, which in turn activates MAPK. The kinases of these signaling cascades are highly conserved, and homologs exist in yeast, Drosophila, and mammalian cells. MEKK1 can phosphorylate and activate MAPKK 1 and MAPKK 2 (MEK1/MEK2) which leads to phosphorylation of MAP kinases. It is also a highly efficient activator of the JNK cascade. The protein contains a putative 1 RING-type zinc finger and 1 SWIM-type zinc finger.Spezifikation
MAP3K1 | |
Polyclonal | |
Unconjugated | |
MAP3K1 | |
MAP3K1, MAPK/ERK kinase kinase 1, MAPKKK1, MEK kinase 1, MEKK, MEKK 1, MEKK1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116667, 26401, 4214 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P53349, Q13233, Q62925 | |
MAP3K1 | |
Peptide | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |