Learn More
Sartorius Manual Mass Comparator
GREENER_CHOICE
Artikelnummer. 30216650 Alle ansehen Sartorius Produkte
Click to view available options
Fassungsvermögen:
32 g
61 g
111 g
211 g
610 g
1110 g
Lesbarkeit:
0,001 mg
0,01 mg
0,1 mg
Packungsgröße:
1 Stück
Artikelnummer. 30216650
Marke: Sartorius MCA66MC
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Dieser Artikel erfordert eine Preisaktualisierung, bevor er bestellt werden kann.
Fordern Sie hier einen Preis für diesen Artikel an.
The new Manual Mass Comparators were designed to improve the usability of our proven Mass Comparators while increasing performance and accuracy. The result is instruments that combine cutting edge metrological specifications with state-of-the-art software.
  • Cubis™ II MCA color touch screen for fast and simple configuration of parameters and workflows.
  • Integrated calibration workflows for ABA, ABBA, AB, BnA cycles to ensure efficient, error-free mass comparison.
  • Fully integrated function for determining the measurement uncertainty in accordance with OIML and ASTM recommendations.
  • Filters for optimal adaptation of the mass comparator to ambient conditions.
  • Additional applications for density determination, statistics and individual identifiers are integrated as standard programs.
  • Automatic Motorized Leveling.
  • Easy logging of reference weight data.
  • Continuous weighing range display: any weight between 0 g and the maximum capacity can be displayed
TRUSTED_SUSTAINABILITY
Spezifikation
Fassungsvermögen 61 g
Displaytyp Color Touchscreen
Lesbarkeit 0,001 mg
Typ Comparator