Learn More
Manganese(II) iodide, ultra dry, 99.99% (metals basis), Thermo Scientific™
Marke: Thermo Scientific Alfa Aesar 047166.06
| Menge | 5g |
|---|
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Ampouled under argon
Chemische Identifikatoren
| 7790-33-2 | |
| MFCD00016225 |
Spezifikation
| Manganese(II) iodide | |
| 7790-33-2 | |
| (metals basis) | |
| 14,5732 | |
| 308.75 | |
| Odorless | |
| Air, light, and moisture sensitive |
| Ultra dry | |
| 5g | |
| MFCD00016225 | |
| Soluble in water. | |
| 99.99% | |
| Ampule | |
| 5.0 g/mL |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.