Learn More
Mangafodipir Trisodium (>85%), TRC
Artikelnummer. 30137583
Change view
Click to view available options
Menge:
10 mg
25 mg
Packungsgröße:
10 Milligramm
25 Milligramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30137583 25 mg 25 Milligramm
17933285 10 mg 10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137583 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCM16370025MG
om dit product te kopen Registreer vandaag om een webaccount aan te maken
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 140678-14-4
Summenformel C22 H27 Mn N4 O14 P2 . 3 Na
Molekulargewicht (g/mol) 757.32
Synonym Manganate(6-), [[rel-[N(R),N'(R)]-N,N'-1,2-ethanediylbis[N-[[3-(hydroxy-κO)-2-methyl-5-[(phosphonooxy)methyl]-4-pyridinyl]methyl]glycinato-κN,κO]](8-)]-, sodium hydrogen (1:3:3), (OC-6-13)-, Manganate(6-), [[N,N'-1,2-ethanediylbis[N-[[3-hydroxy-2-methyl-5-[(phosphonooxy)methyl]-4-pyridinyl]methyl]glycinato]](8-)]-, trisodium trihydrogen, (OC-6-13)-, Manganate(6-), [[rel-[N(R),N'(R)]-N,N'-1,2-ethanediylbis[N-[[3-(hydroxy-κO)-2-methyl-5-[(phosphonooxy)methyl]-4-pyridinyl]methyl]glycinato-κN,κO]](8-)]-, trisodium trihydrogen, (OC-6-13)- (9CI), Mangafodipir trisodium, MnDPDP, S 095, Teslascan, Win 59010
SMILES [Na+].[Na+].[Na+].Cc1ncc(COP(=O)(O)O)c2CN34CCN56CC(=O)[O-][Mn+2]35([O-]C(=O)C4)([O-]c12)[O-]c7c(C)ncc(COP(=O)(O)[O-])c7C6
Spezifikation
Chemischer Name oder Material Mangafodipir Trisodium
Menge 25 mg
InChI-Formel InChI=1S/C22H32N4O14P2.Mn.3Na/c1-13-21(31)17(15(5-23-13)11-39-41(33,34)35)7-25(9-19(27)28)3-4-26(10-20(29)30)8-18-16(12-40-42(36,37)38)6-24-14(2)22(18)32;;;;/h5-6,31-32H,3-4,7-12H2,1-2H3,(H,27,28)(H,29,30)(H2,33,34,35)(H2,36,37,38);;;;/q;+2;3*+1/p-5
Formelmasse 757.0073