Learn More
Value Plastics Male Luer Slip to Plug Adapters
Alle ansehen Cole-Parmer Produkte
Click to view available options
Material (Dichtung):
Nylon
Polypropylen
Packungsgröße:
25 Stück
Beschreibung
Luer fittings allow for a simple yet secure connection. Ideal for capping off fluid transfer system. Quickly connect and disconnect with slip luer design
Spezifikation
Spezifikation
| Zertifizierung/Konformität | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Einlass | Male Luer Slip, Plug, Closed at Grip |
| Material (Dichtung) | Polypropylen |