Learn More
Value Plastics Male Luer Integral Lock Ring to NPT(M) Thread Adapters
Alle ansehen Cole-Parmer Produkte
Click to view available options
Gewindegröße:
1/4-18 NPT(M)
1/8-27 NPT(M)
Material (Dichtung):
Nylon
Polypropylen
Packungsgröße:
25 Stück
Beschreibung
Easily transition from a luer to thread fitting. Luer fittings allow for a simple yet secure twist-lock connection. NPT thread fittings are designed to create a tight seal under pressure
Spezifikation
Spezifikation
| Zertifizierung/Konformität | Non-DEHP;Non-Phthalate;RoHS |
| Gewindegröße | 1/4-18 NPT(M) |
| Material (Dichtung) | Nylon |
| Produkttyp | Male Luer Integral Lock Ring Hex |