Learn More
Melford Magenta-GlcA
Click to view available options
Menge:
100 mg
1 g
Packungsgröße:
1 Gramm
100 Milligramm
Beschreibung
Chromogenic substrate for B-D-Glucuronidase (GUS).
Chromogenic substrate for B-D-Glucuronidase (GUS). Produces a magenta color for (GUS) gene detection.
Spezifikation
Spezifikation
| Menge | 100 mg |