missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
248.00 CHF - 580.00 CHF
Spezifikation
| Antigen | Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 |
|---|---|
| Verdünnung | Western Blot 1 to 2 μg/mL, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 2 to 5 μg/mL, Immunohistochemistry-Paraffin 1:200 |
| Anwendungen | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18388661
|
Bio-Techne
NBP3-06855 |
0.1 mg |
580.00 CHF
0.10 Milligramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18635377
|
Novus Biologicals
NBP3-06855SS |
0.025 mg |
248.00 CHF
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Polyclonal antibody specifically detects Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Spezifikation
| Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Chromatin Research, Transcription Factors and Regulators | |
| PBS | |
| 23135 | |
| IgG | |
| Immunogen affinity purified |
| Western Blot 1 to 2 μg/mL, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 2 to 5 μg/mL, Immunohistochemistry-Paraffin 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 1.14.11, EC 1.14.11.-, JmjC domain-containing protein 3, JMJD3lysine-specific demethylase 6B, jumonji domain containing 3, histone lysine demethylase, Jumonji domain-containing protein 3, KIAA0346jumonji domain containing 3, lysine (K)-specific demethylase 6B, Lysine demethylase 6B | |
| Partial recombinant protein made to an internal portion of the human JMJD3 protein (amino acids 1520-1682) [UniProt O15054] | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts