missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6G Rat, Qfluor 710, Clone: RB6-8C5, enQuireBio™
Rat Monoclonal Antibody
Marke: EnquireBio QAB77-QF710-25ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
Ly-6G | |
Monoclonal | |
0.2mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Ly6c1 | |
Protein A/G Purified | |
RUO | |
17067 | |
IgG2b κ |
Flow Cytometry | |
RB6-8C5 | |
Qfluor 710 | |
NP_001238984 | |
Rat | |
25 μg | |
Primary | |
Store at 2-4°C. |