missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ly-108 Recombinant Rabbit Monoclonal Antibody (4R6M4)

Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA551526
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Ly-108 Recombinant Monoclonal Antibody for Western Blot, IHC (P), ELISA
Ly108, the mouse homologue of human SLAMF6 (signaling lymphocyte activation molecule family member 6), is expressed on NK cells, B cells, and T cells. Engagement of the Ly108 receptor enhances NK cell activity and increases the expression of IFN-gamma. This receptor plays a role in the inability of NK cells to kill Epstein Barr virus-infected B cells in X-linked lymphoproliferative disease. In Ly108 knock-out mice, CD4+ T cells show a diminished ability to produce IL-4 and exhibit defective neutrophil functions, indicating the importance of Ly108 in immune regulation and cellular function. Overall, Ly108 is crucial for modulating immune responses and maintaining effective immune cell activity.
Spezifikation
| Ly-108 | |
| Recombinant Monoclonal | |
| 0.78 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q96DU3, Q9ET39 | |
| SLAMF6 | |
| Recombinant protein of human CD352/SLAMF6. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 4R6M4 | |
| Unconjugated | |
| SLAMF6 | |
| Activating NK receptor; CD352; KAL1; KAL1b; KALI; KALIb; Ly108; Lymphocyte antigen 108; natural killer-, T- and B-cell antigen; NK-T-B-antigen; NTBA; NTB-A; NTBA receptor; RGD1561848; SF2000; SLAM family member 6; Slamf6; UNQ6123/PRO20080 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 114836, 30925, 498287 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur