Learn More
Ly-108 Monoclonal Antibody (eBio13G3-19D (13G3-19D)), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
109.00 CHF - 357.00 CHF
Spezifikation
Antigen | Ly-108 |
---|---|
Klon | eBio13G3-19D (13G3-19D) |
Konzentration | 0.5 mg/mL |
Anwendungen | Flow Cytometry, Immunoprecipitation, Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15247247
|
Affymetrix eBioscience
14-1508-82 |
100 μg |
357.00 CHF
100 Mikrogramm |
Verfügbar ab: 27-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15237247
|
Affymetrix eBioscience
14-1508-80 |
25 μg |
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11. |
||||||
Beschreibung
Description: The eBio13G3-19D monoclonal antibody reacts with mouse Ly108, the mouse homologue of human SLAMF6 (signaling lymphocyte activation molecule family member 6). Mouse Ly108 is also known as NTB-A (NK cell, T cell, B cell antigen), and is expressed on NK, T cells, and B cells. Engagement of the Ly108 receptor enhances NK cell activity and expression of IFN-gamma, and contributes to the inability of NK cells to kill Epstein Barr virus-infected B cells in X-linked lymphoproliferative disease. CD4+ T cells from Ly108 knock-out mice exhibit diminished ability to produce IL-4 and defective neutrophil functions. Staining has not revealed any strain differences between C57BL/6 and BALB/c mice. Other strains have not been tested. Applications Reported: This eBio13G3-19D (13G3-19D) antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunoblotting (WB). Applications Tested: This eBio13G3-19D (13G3-19D) antibody has been tested by flow cytometric analysis of mouse splenocytes and thymocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Ly108 is the mouse homologue of human SLAMF6 (signaling lymphocyte activation molecule family member 6). Mouse Ly108 is expressed on NK, B cells, and T cells. Activation of Ly108 receptor enhances NK cell activity and IFN-gamma expression.Spezifikation
Ly-108 | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
TBS with 0.09% sodium azide; pH 8.0 | |
Activating NK receptor; CD352; KAL1; KAL1b; KALI; KALIb; Ly108; Lymphocyte antigen 108; natural killer-, T- and B-cell antigen; NK-T-B-antigen; NTBA; NTB-A; NTBA receptor; RGD1561848; SF2000; SLAM family member 6; Slamf6; UNQ6123/PRO20080 | |
SLAMF6 | |
Primary | |
4° C | |
SLAMF6 |
eBio13G3-19D (13G3-19D) | |
Flow Cytometry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Mouse | |
Q9ET39 | |
30925 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.