Learn More
Invitrogen™ LXR-a Recombinant Human Protein, Ligand Binding Domain, GST-Tagged
LXR-a (Liver X Receptor alpha) ligand binding domain is a 55.9 kDa recombinant human protein (amino acids 207-447) expressed as a GST fusion protein in baculovirus-infected insect cells.
Marke:  Invitrogen™ PV4657
Additional Details : Gewicht : 0.50000kg
Artikelnummer. 10600185
  • 2455.00 CHF / 50 Mikrogramm
Verfügbar ab: 03-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
LXR alpha was expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator displacement studies.
Spezifikation
Spezifikation
Nuclear Receptor Assay
50 μg
RUO - research use only
Liver X Receptor Alpha
Recombinant
Hormone Receptors
10062
Store in ultra-cold freezer (-68 to -85°C).
Recombinant, Ligand Binding Domain
Human
GST Tag