Learn More
Invitrogen™ LRRK2 [I2020T] Human Protein
LRRK2 is a multidomain protein that includes a kinase domain, a GTPase domain, and a leucine-rich repeat (LRR) domain.
Marke: Invitrogen™ PV5855
Additional Details : Gewicht : 0.01000kg
Beschreibung
Mutations in the LRRK2 gene have been associated with Parkinson 's disease, including the I2020T mutation in the kinase catalytic domain.
Spezifikation
NP_940980.2 | |
204.9kDa | |
Kinase proteins are shipped on dry ice and should be stored at -80°C. | |
Catalytic | |
LRRK Kinase Family | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
1mg | |
RUO - research use only | |
LRRK2 I2020T | |
Human | |
Baculovirus |