missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC8D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11537-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This LRRC8D antibody does not cross-react with LRRC8A.
Component of the volume-regulated anion channel (VRAC, also named VSOAC channel), an anion channel required to maintain a constant cell volume in response to extracellular or intracellular osmotic changes. The VRAC channel conducts iodide better than chloride and may also conduct organic osmolytes like taurine. It is unclear whether LRRC8D constitutes a pore-forming subunit or whether it is closely associated with the pore.Spezifikation
LRRC8D | |
Polyclonal | |
Unconjugated | |
LRRC8D | |
LRRC5, LRRC8D | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
231549, 305131, 55144 | |
Antibody | |
IgG |
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5U308, Q7L1W4, Q8BGR2 | |
Lrrc8d | |
LRRC8D Fusion Protein Ag2138 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Liquid |