missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPXN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11307-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The product encoded by this gene is preferentially expressed in hematopoietic cells and belongs to the paxillin protein family. Similar to other members of this focal-adhesion-associated adaptor-protein family, it has four leucine-rich LD-motifs in the N-terminus and four LIM domains in the C-terminus. It may function in cell type-specific signaling by associating with PYK2, a member of focal adhesion kinase family. As a substrate for a tyrosine kinase in lymphoid cells, this protein may also function in, and be regulated by, tyrosine kinase activity. Alternative splicing results in multiple transcript variants encoding distinct isoforms.Spezifikation
LPXN | |
Polyclonal | |
Unconjugated | |
LPXN | |
LDLP, LDPL, leupaxin, LPXN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
107321, 293783, 9404 | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.42 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O60711, Q99N69 | |
LPXN | |
LPXN Fusion Protein Ag1845 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |