missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPIN2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19545-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Mouse studies suggest that this gene functions during normal adipose tissue development and may play a role in human triglyceride metabolism. This gene represents a candidate gene for human lipodystrophy, characterized by loss of body fat, fatty liver, hypertriglyceridemia, and insulin resistance.Spezifikation
LPIN2 | |
Polyclonal | |
Unconjugated | |
LPIN2 | |
KIAA0249, Lipin 2, LPIN2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9663 | |
-20°C | |
Liquid |
Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q92539 | |
LPIN2 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |