Learn More
Fluid Metering™ VERBINDERSET FUER GERINGE LEISTUNG LEISTUNG
Artikelnummer. 11974138
Encompass
Change view
Click to view available options
Flussrate:
50 ml/min
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Flussrate
11974138 50 ml/min
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 11974138 Lieferant Fluid Metering™ Lieferanten-Nr. R479
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Spezifikation
Flussrate 50 ml/min
Zur Verwendung mit (Anwendung) Low flow (below 50 mL/min.) applications where minimum system dead volume and maximum chemical inertness is required
Zur Verwendung mit Stainless Steel Pump Heads (Except San)
Enthält Four ferrules, two adapters and assembly/removal tools